• USA Home
  • F2641 - Ferrocenemonocarboxylic acid

EMAIL THIS PAGE TO A FRIEND
F2641 Sigma

Ferrocenemonocarboxylic acid

powder

Synonym: (Carboxycyclopentadienyl)cyclopentadienyliron

Purchase

Properties

form   powder
color   brown

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

106887 Ferrocenecarboxylic acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?