• USA Home
  • F2674 - Fraser enrichment supplement

EMAIL THIS PAGE TO A FRIEND
F2674 Sigma

Fraser enrichment supplement

for microbiology

Purchase

Properties

Related Categories Media, Microbiology, Supplements More...
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
shipped in   wet ice
storage temp.   2-8°C

Description

Components

Composition: (per vial)
Acriflavine hydrochloride: 25.00 mg
Nalidixic acid: 4.00 mg

Application

An antimicrobial supplement recommended for isolation, cultivation and enrichment of Listeria monocytogenes from foods and environmental specimens.

Reconstitution

Sufficient for 1 liter medium

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
Protocols & Articles

Articles

Listeria a Survivor

Listeriosis is a serious infection caused by Listeria monocytogenes. In recent years it has been recognized that Listeria is an important public health problem. The disease affects primarily people o...
Jvo Siegrist
Microbiology Focus Edition 2.2
Keywords: AGE, Antibiotics, Diseases, Fermentation, Food Safety, Gastrointestinal, Indicators, Microbiology, Nutrition, Purification

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?