• USA Home
  • F2774 - Kodak® direct exposure film
EMAIL THIS PAGE TO A FRIEND
F2774 Sigma
Kodak® direct exposure film
size 13 cm × 18 cm
Purchase
Properties
mfr. no.   Kodak 154 2505
  DEF-5
packaging   pkg of 25 sheets
size   13 cm × 18 cm
Description
General description
Sheets of film are interleaved with sheets of paper
Coated with emulsion on both sides of clear base, with gelatin coating for abrasion resistance. Recommended for direct exposure to x-rays, high-energy β-emitters, and other ionizing radiation. Excellent for x-ray diffraction analysis and 32P or 45Ca autoradiography.
Specifically designed for direct exposure to x-rays, high-energy β-emitters, and other ionizing radiation.
Excellent for x-ray diffraction analysis and 45Ca or 32P autoradiography.
Emulsion on both sides of clear polyester base, with thin gelatin coat for abrasion-resistance.
Legal Information
Kodak is a registered trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?