• USA Home
  • F2896 - Carestream® Kodak® color film
EMAIL THIS PAGE TO A FRIEND
F2896 Sigma
Carestream® Kodak® color film
Vericolor, III (ASA 160)
Purchase
Properties
Related Categories Electrophoresis Photography Film, Electrophoresis Photography Supplies, Electrophoresis Supplies, Labware
packaging   roll of 36 exposures
mfr. no.   Vericolor, III (ASA 160)
storage temp.   2-8°C
Description
General description
Kodak Professional Films provide the highest quality for accurate scientific imaging. While consumer films provide clear, sharp photographs, they are manufactured with a color-balance bias to allow for emulsion maturation in storage. Professional demands for highly accuate color rendition cannot tolerate this bias. Professional film, therefore, is at or near its color-balance optimum when manufactured. The film will stay near this balance when it is stored at or below 55 °F (13 °C) and processed promptly after exposure - before the expiration date printed on the carton.
Legal Information
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?