• USA Home
  • F2927 - 1-(4-Fluorobenzyl)-5-methoxy-2-methylindole-3-acetic acid

EMAIL THIS PAGE TO A FRIEND
F2927 Sigma

1-(4-Fluorobenzyl)-5-methoxy-2-methylindole-3-acetic acid

≥98% (HPLC), solid

  • Empirical Formula (Hill Notation) C19H18NFO3

  • Molecular Weight 327.35

  •  MDL number MFCD03452919

  •  PubChem Substance ID 24278434

Purchase

Description

Biochem/physiol Actions

Putative inhibitor of multidrug resistance-associated protein 1 (MRP1).

Other Notes

N-benzoyl analog of indomethacin

Packaging

5 mg in glass bottle

Price and Availability


DISCOVER Bioactive Small Molecules

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Drug Transport

Protein-based drug transporters are found in most tissues including liver, kidney, intestine, and brain. Because of their complexity and genetic heterogeneity, these proteins are often produced as re...
Robert Gates
Biofiles Volume 6 Article 1
Keywords: Absorption, Anti-inflammatory agents, Antibiotics, Antimicrobials, Apoptosis, Cancer, Catalog, Cytotoxins, Detoxification, Enzyme-linked immunosorbent assay, Gene expression, Genetic, High performance liquid chromatography, Immunohistochemistry, Mass spectrometry, Metabolites, Methods, Phosphorylations, Separation, The Reporter, Transfection, Type, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?