• USA Home
  • F3005 - N-(3-[2-Furyl]acryloyl)-L-tryptophan methyl ester

EMAIL THIS PAGE TO A FRIEND
F3005 Sigma

N-(3-[2-Furyl]acryloyl)-L-tryptophan methyl ester

  • CAS Number 36020-62-9

  • Empirical Formula (Hill Notation) C19H18N2O4

  • Molecular Weight 338.36

  •  MDL number MFCD00037940

Purchase

Properties

InChI Key   ZZVGGCCUMFTDHV-CMDGGOBGSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?