EMAIL THIS PAGE TO A FRIEND
F3127 Sigma

N-Formylglycine

  • CAS Number 2491-15-8

  • Linear Formula HCONHCH2COOH

  • Molecular Weight 103.08

  •  Beilstein Registry Number 1749108

  •  EC Number 219-649-8

  •  MDL number MFCD00037360

Purchase

Properties

InChI Key   UGJBHEZMOKVTIM-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
MC0547000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

47723 N-Formylglycine, ≥98.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?