EMAIL THIS PAGE TO A FRIEND
F3260 Sigma

Fetal Bovine Serum

purified by dialysis (MWCO 1,000 Da), cell culture tested

Synonym: FBS, sera, serum

Purchase

Properties

sterility   sterile-filtered (MWCO 1,000 Da)
purified by   dialysis (MWCO 1,000 Da)
origin   USA origin
impurities   endotoxin, tested
suitability   cell culture tested
shipped in   dry ice
storage temp.   −20°C

Description

Analysis Note

Endotoxin and hemoglobin tested

Preparation Note

Dialyzed by ultrafiltration against 0.15 M NaCl.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

F0392 Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?