• USA Home
  • F3652 - Ferritin–Biotinamidocaproyl Labeled from Equine Spleen

EMAIL THIS PAGE TO A FRIEND
F3652 Sigma

Ferritin–Biotinamidocaproyl Labeled from Equine Spleen

buffered aqueous glycerol solution

Purchase

Properties

form   buffered aqueous glycerol solution
composition   Protein, ~10 mg/mL BCA
extent of labeling   5-10 mol biotin per mol apoferritin
storage temp.   −20°C

Description

Other Notes

holo-Ferritin coupled by amide bond to biotin through an aminocaproyl spacer

Packaging

Package size based on protein content

Physical form

Solution in 50% glycerol containing 0.15 M sodium chloride, 0.01 M sodium phosphate, pH 7.2 and 0.02% sodium azide

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?