• USA Home
  • F3755 - 3-(2-Furyl)acrylic acid

EMAIL THIS PAGE TO A FRIEND
F3755 Sigma

3-(2-Furyl)acrylic acid

Purchase

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
LT8560000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?