• USA Home
  • F3801 - N-Formyl-D-phenylglycine trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
F3801 Sigma

N-Formyl-D-phenylglycine trifluoroacetate salt

>99% (TLC)

  • Empirical Formula (Hill Notation) C9H9NO3

  • Molecular Weight 179.17

  •  MDL number MFCD03452921

  •  PubChem Substance ID 329799705

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   PKLCDQYRDFFAJJ-DDWIOCJRSA-N
assay   >99% (TLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?