• USA Home
  • F3802 - Fluorescein-β-D-glucopyranoside polyacrylamide

EMAIL THIS PAGE TO A FRIEND
F3802 Sigma

Fluorescein-β-D-glucopyranoside polyacrylamide

powder

Synonym: β-D-Glc-PAA-FI

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates D-F, Polyacrylamide (PAA)-based Glycoconjugates More...
form   powder
mol wt   mol wt ~30 kDa
color   yellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?