• USA Home
  • F3880 - N-(3-Fluoranthyl)maleimide

EMAIL THIS PAGE TO A FRIEND
F3880 Sigma

N-(3-Fluoranthyl)maleimide

Synonym: FAM

  • CAS Number 60354-76-9

  • Empirical Formula (Hill Notation) C20H11NO2

  • Molecular Weight 297.31

  •  Beilstein Registry Number 1483865

  •  EC Number 262-197-1

  •  MDL number MFCD00042618

  •  PubChem Substance ID 24894797

Purchase

Description

Application

Fluorescent protein marker

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?