• USA Home
  • F3899 - Monoclonal Anti-CD19−FITC antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
F3899 Sigma

Monoclonal Anti-CD19−FITC antibody produced in mouse

clone SJ25-C1, purified immunoglobulin, buffered aqueous solution

Synonym: Monoclonal Anti-CD19

Purchase

Properties

Related Categories Alphabetical Index, Analytical Cytology, Antibodies, Antibodies for Cell Biology, Antibodies for Immune Cell Signaling and Blood,
application(s)   flow cytometry: 10 μL using 1 × 106 cells
clone   SJ25-C1, monoclonal
antibody form   purified immunoglobulin
form   buffered aqueous solution
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... CD19(930)
biological source   mouse
conjugate   FITC conjugate

Description

Immunogen

NALM-1 human cell line.

Antigen Background

CD19 is a member of the immunoglobulin superfamily and has two Ig-like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of κ or λ light chains. It is expressed on approximately 10% of normal human peripheral blood cells and approximately 60% of splenic lymphocytes. It is not expressed on granulocytes, monocytes or T cells as defined by CD3 expression.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Specificity

Recognizes the human CD19 (90 kDa) integral membrane glycoprotein.
3rd Workshop: code no. 073

Preparation Note

Prepared by conjugation to fluorescein isothiocyanate isomer I (FITC). This green dye is efficiently excited at 495 nm and emits at 525 nm.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?