• USA Home
  • F4026 - Fibrinogen from rabbit plasma

EMAIL THIS PAGE TO A FRIEND
F4026 Sigma

Fibrinogen from rabbit plasma

Purchase

Properties

composition   Protein, ~60% (over 85% of protein clottable)
storage temp.   −20°C

Description

Analysis Note

Protein determined by biuret.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?