• USA Home
  • F4029 - Ferredoxin from Porphyra umbilicalis (red marine algae)

EMAIL THIS PAGE TO A FRIEND
F4029 Sigma

Ferredoxin from Porphyra umbilicalis (red marine algae)

lyophilized powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Proteins and Derivatives, Redox More...
form   lyophilized powder
purified by   chromatography
storage temp.   2-8°C

Description

Biochem/physiol Actions

Ferredoxins are iron-containing proteins that serve as electron-transfer catalysts in photosynthesis.

Packaging

Package sizes are based on ferredoxin content.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?