• USA Home
  • F4391 - p-Fluoro-D-phenylalanine

EMAIL THIS PAGE TO A FRIEND
F4391 Sigma

p-Fluoro-D-phenylalanine

Synonym: 4-Fluoro-D-phenylalanine

  • CAS Number 18125-46-7

  • Empirical Formula (Hill Notation) C9H10FNO2

  • Molecular Weight 183.18

  •  Beilstein Registry Number 2416149

  •  MDL number MFCD00063063

  •  PubChem Substance ID 24894862

Purchase

Description

Biochem/physiol Actions

Inactive isomer of p-fluorophenylalanine

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?