• USA Home
  • F4677 - Sartorius Proline® XL electronic pipetting aide
EMAIL THIS PAGE TO A FRIEND
F4677 Sigma
Sartorius Proline® XL electronic pipetting aide
Replacement filter
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Manufacturer Browser, Pipette Controllers and Pumps, Sartorius Biohit,
sterility   non-sterile
mfr. no.   Sartorius, 712912
Description
General description
Simply select a volume to multidispense using your choice of any common graduated or non-graduated glass and plastic pipettes (adapters are included). Sartorius Proline XL of the Biohitfamily performs electronically controlled pipetting/dispensing of volumes between 0.1-25 mL. The standard 5 mL and 10 mL tips fit the XL ensuring, thus, optimum versatility. Features include: a pipettor and a dispenser; Different kinds of tips/pipettes can be used by simply changing the adapter; No need for visual control; Both left and right handed; Can be used both as a microprocessor-controlled or manually-operated pipetting controller; Can be programmed for a certain volume range to prevent overflow with small pipettes/tips; Hydrophobic autoclavable filter prevents overaspiration
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?