• USA Home
  • F4756 - Fumarase from rabbit liver

EMAIL THIS PAGE TO A FRIEND
F4756 Sigma

Fumarase from rabbit liver

ammonium sulfate suspension, 200-500 units/mg protein (Lowry)

Synonym: L-Malate hydrolyase, Fumarate hydratase

Purchase

Description

Unit Definition

One unit will convert 1.0 μmole of L-malate to fumarate per min at pH 7.6 at 25°C.

Physical form

Suspension in 3.2 M (NH4)2SO4, pH 7.5

Price and Availability


DISCOVER Bioactive Small Molecules

All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Protocols

Enzymatic Assay of Fumarase

To standardize a procedure for the enzymatic assay of Fumarase in Analytical Services at Sigma-Aldrich - St. Louis.
Keywords: Extinction coefficient

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

F1757 Fumarase from porcine heart, ammonium sulfate suspension, 300-500 units/mg protein (biuret)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?