EMAIL THIS PAGE TO A FRIEND
F4758 Sigma

Fluorinert FC-77

Purchase

Properties

Related Categories Cell Biology, Density Gradient Media, Hematology and Histology, Miscellaneous
bp   ~100 °C(lit.)
density   1.78 g/mL at 25 °C(lit.)

Description

Application

For possible use as a high density chase solution in density gradient studies.

Other Notes

A trademark for a liquid mixture of completely fluorinated aliphatic compounds.

Legal Information

Fluorinert is a trademark of 3M Company

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?