EMAIL THIS PAGE TO A FRIEND
F4896 Sigma

Fast Red PDC salt

Synonym: 5-Butylaminosulfonyl-2-methoxybenzenediazonium chloride hemi(zinc chloride) salt

  •  Colour Index Number 37151

  •  EC Number 201-261-5

Purchase

Properties

composition   Dye content, >90%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?