EMAIL THIS PAGE TO A FRIEND
F5003 Sigma

N-Formyl-Met-Val

≥97% (HPLC)

  • CAS Number 29790-45-2

  • Empirical Formula (Hill Notation) C11H20N2O4S

  • Molecular Weight 276.35

  •  MDL number MFCD00037542

  •  PubChem Substance ID 24894874

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chemotactic Peptides, Immunomodulators, Peptides and Proteins,
InChI Key   DWZMYBRYTYGZDG-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

NFor-Met-Val

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?