• USA Home
  • F5129 - Fluorescein dilaurate

EMAIL THIS PAGE TO A FRIEND
F5129 Sigma

Fluorescein dilaurate

  • CAS Number 7308-90-9

  • Empirical Formula (Hill Notation) C44H56O7

  • Molecular Weight 696.91

  •  Beilstein Registry Number 6034843

  •  EC Number 230-764-2

  •  MDL number MFCD00038409

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Substrate, Fatty Acyls, Fluorogen, Lipids More...
InChI Key   MYTRGBGGRICZGN-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

46943 Fluorescein dilaurate, suitable for fluorescence, ≥97.0% (HPCE)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?