EMAIL THIS PAGE TO A FRIEND
F6051 Sigma

Fisonate

≥99% (NMR), solid

  • Empirical Formula (Hill Notation) C12H20F6NO4P

  • Molecular Weight 387.26

  •  MDL number MFCD00285808

Purchase

Description

Biochem/physiol Actions

A broad spectrum inhibitor of serine protease.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?