EMAIL THIS PAGE TO A FRIEND
F6263 Sigma
Film hangers
For 14 x 17 in. and 35 x 43 cm film
Purchase
Description
General description
For tank development of x-ray film. Stainless steel frame has attached clips to hold the film taut. The holder for 36 in. long film holds the film tightly in a "U" shape to fit a standard tank.
See also: Drying Rack for Film. Storage Rack for Film Hangers
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?