• USA Home
  • F6411 - Millipore® glass-fiber filters, AP series
EMAIL THIS PAGE TO A FRIEND
F6411 Sigma
Millipore® glass-fiber filters, AP series
Type AP 25, circles size 2.5 cm
Purchase
Properties
Related Categories Filtration Supplies, Glass Fiber Filters, Labware, Manufacturer Browser, Millipore,
material   borosilicate with acrylic binder
  circles
packaging   pkg of 100 ea
mfr. no.   Millipore, AP2502500
circles size   2.5 cm
pore size   2.0 μm
Description
General description
AP Series filters are primarily designed as prefilters to retain contaminants that would quickly plug membrane filters.
Suitability
Same retention as Type AP20 but thicker, much higher capacity.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?