• USA Home
  • F6647 - IgG2b−FITC Isotype Control from murine myeloma

EMAIL THIS PAGE TO A FRIEND
F6647 Sigma

IgG2b−FITC Isotype Control from murine myeloma

clone MOPC-141, purified immunoglobulin, buffered aqueous solution

Synonym: IgG2b Isotype Control from murine myeloma, Mouse IgG2b, κ

Purchase

Properties

Related Categories Analytical Cytology, Antibodies, Antibody Characterization and Analysis, Biochemicals and Reagents, Isotype Controls,
biological source   mouse
antibody form   purified immunoglobulin
clone   MOPC-141, monoclonal
form   buffered aqueous solution
storage condition   protect from light
application(s)   flow cytometry: suitable
isotype   IgG2bκ
conjugate   FITC conjugate
shipped in   wet ice
storage temp.   2-8°C

Description

Preparation Note

Prepared by conjugation to fluorescein isothiocyanate isomer I (FITC). This green dye is efficiently excited at 495 nm and emits at 525 nm.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?