• USA Home
  • F6671 - Ferredoxin from Clostridium pasteurianum

EMAIL THIS PAGE TO A FRIEND
F6671 Sigma

Ferredoxin from Clostridium pasteurianum

lyophilized powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Proteins and Derivatives, Redox More...
form   lyophilized powder
composition   ferredoxin, ~30 wt. % E1%/390 = 34
storage temp.   −20°C

Description

Biochem/physiol Actions

Ferredoxins are iron-containing proteins that serve as electron-transfer catalysts in photosynthesis.

Packaging

Package sizes are based on ferredoxin content.

Preparation Note

Isolated by a modified procedure of Rabinowitz, J., Methods in Enzymology, 24, 431 (1972).

Physical form

Lyophilized powder containing Tris buffer

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

F3013 Ferredoxin from Spinacia oleracea (spinach), lyophilized powder, Ferredoxin ≥15 wt. %

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?