• USA Home
  • F6882 - N-Formyl-Met-Leu-p-iodo-Phe

EMAIL THIS PAGE TO A FRIEND
F6882 Sigma

N-Formyl-Met-Leu-p-iodo-Phe

≥85% (HPLC)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chemotactic Peptides, Immunomodulators, Peptides and Proteins,
InChI Key   RDQXFDPVOCUTTL-BZSNNMDCSA-N
assay   ≥85% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

NFor-Met-Leu-pI-Phe

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?