• USA Home
  • F7010 - Filter capsule: pleated membrane

EMAIL THIS PAGE TO A FRIEND
F7010 Sigma

Filter capsule: pleated membrane

0.2 μm medium volume, effective filtration area 1380 cm2 (231 in2), sterile; γ-irradiated

Synonym: membrane filter, membrane filters, pleated filters

Purchase

Properties

sterility   sterile; γ-irradiated
mfr. no.   Gelman 12117
effective filtration area   1380 cm2 (231 in2)

Description

Application

For sterilization of 100-1000L of distilled or DI water, diluents and laboratory reagents or up to 30L of Tissue Culture media (without serum).

Other Notes

Pleated membrane of non-media migrating filter material in a polycarbonate capsule.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?