• USA Home
  • F7037 - Lipopolysaccharide–FITC from Escherichia coli O26:B6

EMAIL THIS PAGE TO A FRIEND
F7037 Sigma

Lipopolysaccharide–FITC from Escherichia coli O26:B6

lyophilized powder

Synonym: Fluorescein isothiocyanate-lipopolysaccharide from Escherichia coli O26:B6

Purchase

Properties

form   lyophilized powder
storage temp.   2-8°C

Description

Preparation Note

Prepared from lipopolysaccharide chromatographically purified by gel filtration

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

F8666 Lipopolysaccharides from Escherichia coli O55:B5, FITC conjugate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?