• USA Home
  • F7128 - Fibrinogen from canine plasma

EMAIL THIS PAGE TO A FRIEND
F7128 Sigma

Fibrinogen from canine plasma

Synonym: Factor I

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Coagulation Proteins and Reagents, Enzymes, Inhibitors, and Substrates, Fibrinogen,
mol wt   α-chain mol wt 63.5 kDa
  β-chain mol wt 56 kDa
  γ chain mol wt 47 kDa (about 4% carbohydrate content)
  soluble dimer mol wt 340 kDa
storage temp.   −20°C
Gene Information   dog ... FGA(475473)

Description

Analysis Note

Protein determined by biuret.

Quality

~55% protein (over 60% of protein clottable)
Contains ~20% sodium citrate and ~25% sodium chloride.

Biochem/physiol Actions

Fibrinogen is an acute phase protein that is part of the coagulation cascade of proteins. The end result of the cascade is the production of thrombin that converts fibrinogen to fibrin. Thrombin rapidly proteolyses fibrinogen, releasing fibrinopeptide A. The loss of this small peptide is not sufficient to make the resulting fibrin molecule insoluble, but it tends to form complexes with adjacent fibrin and fibrinogen molecules. Thrombin then cleaves a second peptide, fibrinopeptide B, from fibrin and the fibrin monomers formed then polymerize spontaneously to form an insoluble gel. The polymerized fibrin is held together by noncovalent and electrostatic forces and stabilized by the transamidating enzyme, factor XIIIa, that is produced by the action of thrombin on factor XIII. The insoluble fibrin aggregates (clots) and aggregated platelets then block the damaged blood vessel and prevent further bleeding. The amount of fibrinogen in the plasma can serve as a nonspecific indicator of whether or not an inflammatory process is present in the body. Fibrinogen from any mammalian source will be cleaved by thrombin from any mammalian source.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

F8630 Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?