EMAIL THIS PAGE TO A FRIEND
F7148 Sigma

syringe filters

pore size 0.22 μm, diam. 13 mm, ethylene oxide treated

Synonym: Millex® syringe filter units, disposable, Durapore® PVDF

Purchase

Properties

Related Categories Filtration Supplies, Labware, Manufacturer Browser, Millex syringe filter units, disposable, Durapore PVDF, Millipore,
material   PVDF membrane (hydrophilic)
  high-density polyethylene housing
sterility   ethylene oxide treated
packaging   pack of 100
mfr. no.   Millipore SLGV013SL
  Millipore, SLGV013SL
diam.   13 mm
volume   ≤10 mL
pore size   0.22 μm
fitting   inlet female Luer (lock)
  outlet male Luer

Description

Application

Tissue culture; sterilization

Ultra-low protein binding (<1 μg/cm2). Component materials meet USP XXI Class VI Biological Tests for plastics. Water extractables <0.5%

General description

Units contain Durapore Hydrophilic PVDF filter membranes in plastic housings with Luer lock inlet and Luer slip outlet fittings.

Units smaller than 25 mm diameter are specifically designed to minimize hold-up in filtration of small volumes. The 3-4 mm units are recommended for volumes of <1 mL; the 13 mm units are recommended for volumes of 0.5 - 10 mL.

Suitability

Sterilization of aqueous solutions

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany

Millex is a registered trademark of Merck KGaA, Darmstadt, Germany

Price and Availability


Pharma QC
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

Sterile Millex® Filters for Cell Culture

Millex syringe filters are ideal for sterilizing 1 mL – 200 mL of cell culture reagents, such as cell culture media, antibiotics, supplements, sera, viruses for infection, and other biological soluti...
Keywords: Antibiotics, Capillary electrophoresis, Cell culture, Culture media, Filtration, Growth factors, Pharmaceutical, Sterilizations

Protocols

TRI Reagent® Protocol

1A. Tissue: Homogenize tissue samples in TRI Reagent (1 ml per 50–100 mg of tissue) in a Polytron® or other appropriate homogenizer.
Keywords: AGE, Buffers, Cell disruption, Centrifugation, Degradations, Digestions, Electrophoresis, Evaporation, Gel electrophoresis, Homogenization, PAGE, Polymerase chain reaction, Precipitation, Sample preparations, Separation, Solvents, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?