• USA Home
  • F7161 - Millipore® glass-fiber filters
EMAIL THIS PAGE TO A FRIEND
F7161 Sigma
Millipore® glass-fiber filters
Type 2, retention 1.0 μm, circles size 4.7 cm
Purchase
Properties
Related Categories Filtration Supplies, Glass Fiber Filters, Labware, Manufacturer Browser, Millipore,
material   borosilicate glass
  circles
packaging   pkg of 100 ea
mfr. no.   Millipore, APFB04700
circles size   4.7 cm
pore size   1.0 μm retention
Description
Application
Highly absorbent, retentive. For biological fluids and biopolymers
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?