• USA Home
  • F7260 - Filtration kit replacement filters
EMAIL THIS PAGE TO A FRIEND
F7260 Sigma
Filtration kit replacement filters
filter size 0.8 μm × 0.45 μm, Sterile
Synonym: filtration kit, replacement filter
Purchase
Properties
Related Categories Filtration Supplies, Labware, Membrane Filters, Polyethersufone Membrane Filters
sterility   sterile; γ-irradiated
feature   effective filtration area 500 cm2 78 in2
filter size   0.8 μm × 0.45 μm
Description
General description
Disposable ready-to-use replacement filter capsules for the Liquid Media Filtration Kit (FK-1) to afford flexibility for your applications. All filter capsule materials have been evaluated for biosafety in accordance with U.S.P. (United States Pharmacopoeia) Class VI Plastics Tests to ensure safety of materials. All filters described are intended for use in positive pressure filtration systems at pressures up to 50 psi. Individual filters or serial combinations of filters can be used to pre-filter and/or sterilize Tissue Culture media (without serum) or reagents in the volumes indicated. Individual filter efficiency and throughput will vary with the product being filtered. Products such as serum, medium containing serum, or contaminated liquids will reduce the capacity of the filter
Two layers of non-media migrating filter material in a polycarbonate capsule.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?