• USA Home
  • F7265 - Fendiline hydrochloride

EMAIL THIS PAGE TO A FRIEND
F7265 Sigma

Fendiline hydrochloride

Synonym: N-(3,3-Diphenylpropyl)-α-methylbenzylamine

Purchase

Description

Biochem/physiol Actions

Coronary vasodilator

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DP4790000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?