• USA Home
  • F7272 - Carestream® Kodak® filter
EMAIL THIS PAGE TO A FRIEND
F7272 Sigma
Carestream® Kodak® filter
Wratten 22
Purchase
Properties
Related Categories Autoradiography, Autoradiography Film Accessories, Carestream Kodak, Carestream Kodak Photography Supplies, Carestream Kodak filter,
material   gelatin filters 75mm (3 in) square
mfr. no.   Kodak
size   75 mm (3 in.) × 75 mm
Description
Features and Benefits
Deep orange filter provides optimal contrast for black-and-white photography of ethidium bromide stained nucleic acids. A UV filter such as Wratten 2A (above), used between the light source and the Wratten 22, reduces background and protects the Wratten 22 against photo-bleaching.
General description
Primarily for black-and-white photography intended to vary contrast and tonal reproduction.
Legal Information
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?