• USA Home
  • F7386 - 3-Fluoro-3-deoxy-D-glucose

EMAIL THIS PAGE TO A FRIEND
F7386 Sigma

3-Fluoro-3-deoxy-D-glucose

Synonym: 3-Deoxy-3-fluoro-D-glucose

  • CAS Number 14049-03-7

  • Empirical Formula (Hill Notation) C6H11FO5

  • Molecular Weight 182.15

  •  Beilstein Registry Number 1865171

  •  MDL number MFCD00063302

  •  PubChem Substance ID 24894941

Purchase

Properties

Related Categories Activity, Aldolase reductase, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates,
InChI Key   BUMRBAMACDBPKO-CBPJZXOFSA-N
mp   115-117 °C(lit.)
storage temp.   2-8°C

Description

Packaging

50 mg in glass bottle

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?