• USA Home
  • F7753 - α-L-Fucosidase solution from bovine epididymis

EMAIL THIS PAGE TO A FRIEND
F7753 Sigma

α-L-Fucosidase solution from bovine epididymis

ammonium sulfate suspension, ~2 units/mg protein (modified Warburg-Christian)

Synonym: α-L-Fucoside fucohydrolase

Purchase

Properties

Related Categories 3.2.x.x Glycosidases, 3.x.x.x Hydrolases, Biochemicals and Reagents, Enzyme Class Index, Enzymes, Inhibitors, and Substrates More...
form   ammonium sulfate suspension
foreign activity   β-N-acetylglucosaminidase <1.5%
storage temp.   2-8°C

Description

Other Notes

May be similar to the rat epididymal enzyme reported by Carlsen, R.B. and Pierce, J.G., J. Biol. Chem., 247, 23 (1972).

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl α-L-fucoside to p-nitrophenol and L-fucose per min at pH 6.5 at 25°C.

Physical form

Suspension in 2.5 M (NH4)2SO4, pH 5.8

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

F5884 α-L-Fucosidase from bovine kidney, ammonium sulfate suspension, ≥2.0 units/mg protein (biuret)

substrate

Product #

Description

Add to Cart

N3628 4-Nitrophenyl α-L-fuco­pyran­oside, ≥98% (TLC)
M8527 4-Methylumbelliferyl α-L-fucopyranoside, ≥97% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?