• USA Home
  • F8175 - Farnesylthiosalicylic acid

EMAIL THIS PAGE TO A FRIEND
F8175 Sigma

Farnesylthiosalicylic acid

>98% (HPLC), solid

Synonym: FTS, S-trans,trans-Farnesylthiosalicylic acid, [3,7,11-Trimethyldodeca-2,6,10-trienyl]-2-thiobenzoic acid

Purchase

Properties

InChI Key   WUILNKCFCLNXOK-CFBAGHHKSA-N
assay   >98% (HPLC)
form   solid
color   off-white
shipped in   wet ice
storage temp.   2-8°C

Description

Biochem/physiol Actions

Specific, non-toxic Ras antagonist that dislodges Ras from its membrane anchorage domains and accelerates its degradation

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?