• USA Home
  • F8398 - Millex® syringe filter units, disposable, mixed esters
EMAIL THIS PAGE TO A FRIEND
F8398 Sigma
Millex® syringe filter units, disposable, mixed esters
pore size 0.8 μm, diam. 33 mm
  •  eCl@ss 32070500
Purchase
Properties
Related Categories Filtration Supplies, Labware, Manufacturer Browser, Millex syringe filter units, disposable, mixed esters, Millipore,
material   mixed esters membrane
  modified acrylic housing
sterility   sterile; ethylene oxide treated
packaging   pack of 50 ea
mfr. no.   Millipore SLAA033
  Millipore, SLAA033SS
diam.   33 mm
volume   ≤100 mL
pore size   0.8 μm
fitting   inlet female Luer (lock)
  outlet male Luer
Description
Application
Prefiltration; particulate removal
General description
Units contain mixed cellulose ester filter membranes in plastic housings with Luer lock inlet and Luer slip outlet fittings. The large filter area reduces the pressure required to empty syringe and increases flow rate.
Use for clarification and pre-filtration of water, buffers and other aqueous solutions.
Suitability
Suitable with aqueous solutions.
Legal Information
Millex is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?