• USA Home
  • F8416 - mouse igg1-FITC/igg1-Quantum Red dual-tag isotype control

EMAIL THIS PAGE TO A FRIEND
F8416 Sigma

mouse igg1-FITC/igg1-Quantum Red dual-tag isotype control

200 μg/mL, whole antiserum, buffered aqueous solution

Purchase

Properties

antibody form   whole antiserum
form   buffered aqueous solution
packaging   vial of 100 μg
concentration   200 μg/mL
conjugate   FITC conjugate
  Quantum Red® conjugate
shipped in   wet ice
storage temp.   2-8°C

Description

Physical form

Solution in 0.01 M phosphate buffered saline (PBS), pH 7.4, containing 1% BSA and 15 mM sodium azide as preservative

Legal Information

DUAL-TAG is a trademark of Sigma-Aldrich Co. LLC

Quantum Red is a trademark of Sigma-Aldrich Co. LLC

Texas Red is a registered trademark of Life Technologies

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?