• USA Home
  • F8551 - Fractalkine, Chemokine Domain from rat

EMAIL THIS PAGE TO A FRIEND
F8551 Sigma

Fractalkine, Chemokine Domain from rat

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder

Synonym: CX3CL-1, Neurotactin

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chemokines, Cytokines and Growth Factors, Cytokines, Growth Factors and Hormones,
recombinant   expressed in E. coli
assay   >97% (SDS-PAGE)
form   lyophilized powder
mol wt   predicted mol wt ~8.8 kDa
impurities   endotoxin, tested
storage temp.   −20°C
Gene Information   rat ... Cx3cl1(89808)

Description

Analysis Note

The biological activity is measured by its ability to induce chemotaxis of human peripheral blood lymphocytes.

Other Notes

Represents a 76 amino acid variant of the chemokine domain.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% TFA containing 1.25 mg bovine serum albumin.

Biochem/physiol Actions

With the exception of the stalk region, rat fractalkine shares approximately 83% sequence identity with human and mouse fractalkine.

Price and Availability

Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?