EMAIL THIS PAGE TO A FRIEND
F8916 Sigma

Fmoc-Arg(Pbf)-OH

Synonym: Nα-Fmoc-Nω-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine, Nα-Fmoc-Nω-Pbf-L-arginine

  • CAS Number 154445-77-9

  • Empirical Formula (Hill Notation) C34H40N4O7S

  • Molecular Weight 648.77

  •  Beilstein Registry Number 8302671

  •  MDL number MFCD00235804

  •  PubChem Substance ID 24894980

Purchase

Properties

Related Categories A - H, Amino Acids, Amino Acids & Derivatives, Arginine, Biochemicals and Reagents,
InChI Key   HNICLNKVURBTKV-NDEPHWFRSA-N
storage temp.   −20°C

Description

Other Notes

This product has been replaced by 47349-ALDRICH | Fmoc-Arg(Pbf)-OH ≥98.0% (HPLC)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?