• USA Home
  • F8926 - Anti-phospho-FAK (pTyr577) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
F8926 Sigma

Anti-phospho-FAK (pTyr577) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-phospho-Focal Adhesion Kinase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, Antibodies to Phosphoproteins at Phosphotyrosine (pTyr),
species reactivity   frog, mouse, human
application(s)   microarray: suitable
  western blot: 1:1,000 using NIH3T3 cells treated with PDGF
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... PTK2(5747)
mouse ... Ptk2(14083)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic phosphopeptide derived from the region of FAK that contains tyrosine 577.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 50% glycerol, 1.0 mg/ml BSA (IgG and protease free) and 0.05% sodium azide.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?