• USA Home
  • F9049 - Anti-Fibroblast Growth Factor-10 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
F9049 Sigma

Anti-Fibroblast Growth Factor-10 antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym: Anti-FGF-10

Purchase

Description

Immunogen

recombinant human fibroblast growth factor 10 expressed in E. coli.

General description

Fibroblast Growth Factor 10 (FGF-10) is a protein expressed mainly in the embryo and adult lung as well as in adult fibroblasts and pre-adipocytes. It has mitogenic activity for epithelial and epidermal cells but not for fibroblasts. FGF-10 plays a crucial role in lung development, wound healing and limb bud formation. It is also required for normal branching morphogenesis. Anti-Fibroblast Growth Factor-10 antibody can be used in indirect ELISA at a working antibody concentration of 0.5-1.0 μg/ml. Goat anti-Fibroblast Growth Factor-10 antibody reacts specifically with recombinant human FGF-10. The antibody exhibits 30% (approx.) cross-reactivity with recombinant human FGF-7 and 15% with recombinant human FGF-5.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Application

Anti-Fibroblast Growth Factor-10 antibody can be used in western blotting at a working antibody concentration of 0.1-0.2 μg/ml.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?