• USA Home
  • F9050 - Anti-Flt-3/Flk-2 ligand (FL) antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
F9050 Sigma

Anti-Flt-3/Flk-2 ligand (FL) antibody produced in goat

affinity isolated antibody, lyophilized powder

Description

Immunogen

recombinant human Flt-3 ligand expressed in Sf 21 cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?