• USA Home
  • F9674 - Fibroblast Growth Factor Receptor-2β (IIIb)/Fc Chimera human

EMAIL THIS PAGE TO A FRIEND
F9674 Sigma

Fibroblast Growth Factor Receptor-2β (IIIb)/Fc Chimera human

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym: FGFR-2β (IIIb)/Fc Chimera

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cytokines and Growth Factors, Cytokines, Growth Factors and Hormones, FGF Receptors,
recombinant   expressed in NSO cells
assay   >90% (SDS-PAGE)
form   lyophilized powder
storage temp.   −20°C
Gene Information   human ... FGFR2(2263)

Description

Analysis Note

The bioactivity is measured by its ability to inhibit human fibroblast growth factor acidic-dependent proliferation of NR6 cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline

Biochem/physiol Actions

FGFR2 (fibroblast growth factor receptor 2) facilitates tumor growth, invasiveness, metastasis and angiogenesis. It plays an essential role in the development of mammary glands, and its expression is elevated in breast cancers. Studies in North Indian population suggest that polymorphisms in this gene might be linked to susceptibility to breast cancer. It is anti-apoptotic in nature, and suppresses p53-mediated apoptosis, by inducing the elevation of MDM2 levels. Mutations in this gene are linked to mild Crouzon syndrome and craniosynostosis.

General description

FGFR2 (fibroblast growth factor receptor 2) is a member of the tyrosine kinase receptor family called FGFR. This gene is localized to human chromosome 10q26, and is composed of 2 exons. Its ligand-binding domain in present in its extracellular region, which is composed of three immunoglobulin (Ig)-like domains. It spans the membrane once, and its tyrosine kinase domain is localized to its intracellular region. Alternative splicing of this gene gives rise to two isoforms- epithelial (FGFR2IIIb) and mesenchymal (FGFR2IIIc).

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?