• USA Home
  • F9758 - N-Formyl-Met-Leu-Phe benzylamide

EMAIL THIS PAGE TO A FRIEND
F9758 Sigma

N-Formyl-Met-Leu-Phe benzylamide

≥97% (HPLC)

  • CAS Number 80180-62-7

  • Empirical Formula (Hill Notation) C28H38N4O4S

  • Molecular Weight 526.69

  •  MDL number MFCD00057573

  •  PubChem Substance ID 24895002

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chemotactic Peptides, Immunomodulators, Peptides and Proteins,
InChI Key   SYWWPBYRTBWRKE-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Nform-Met-Leu-Phe-BzlA

Biochem/physiol Actions

A potent chemotactic peptide

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?