EMAIL THIS PAGE TO A FRIEND
F9801 Sigma

FIM-1 diacetate

  • Empirical Formula (Hill Notation) C49H36N4O10

  • Molecular Weight 840.83

  •  MDL number MFCD00285807

Purchase

Properties

Related Categories Cell Biology, E-F, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
solubility   DMSO or DMF: soluble
shipped in   dry ice
storage temp.   −20°C

Description

Biochem/physiol Actions

Membrane permeable derivative of FIM-1. When taken up by cells, endogenous esterases convert this product to the active FIM-1.

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?